missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Bromo-4-methoxybenzoate 98.0+%, TCI America™

Catalog No. M22645G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M22645G Supplier TCI America Supplier No. M22645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35450-37-4
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.07
MDL Number MFCD00210463
InChI Key ZREVPFANJBZHEU-UHFFFAOYSA-N
Synonym 3-bromo-4-methoxybenzoic acid methyl ester, methyl3-bromo-4-methoxybenzoate, benzoic acid, 3-bromo-4-methoxy-, methyl ester, acmc-209igx, methyl 3-bromo-p-anisate, rarechem al bf 0247, methyl-3-bromo-4-methoxybenzoate, 3-bromo-p-anisic acid methyl ester, #, methyl 3-bromo-4-methyloxy benzoate
PubChem CID 611662
IUPAC Name methyl 3-bromo-4-methoxybenzoate
SMILES COC(=O)C1=CC(Br)=C(OC)C=C1

Specifications

Melting Point 95°C
Color White
Quantity 5 g
Formula Weight 245.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Bromo-4-methoxybenzoate