missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-bromo-4-methoxybenzoate, 98%

Catalog No. AAH2729709
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2729709 10 g
AAH2729703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2729709 Supplier Thermo Scientific Chemicals Supplier No. H2729709
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 35450-37-4 | C9H9BrO3 | 245.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35450-37-4
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.07
MDL Number MFCD00210463
InChI Key ZREVPFANJBZHEU-UHFFFAOYSA-N
Synonym 3-bromo-4-methoxybenzoic acid methyl ester, methyl3-bromo-4-methoxybenzoate, benzoic acid, 3-bromo-4-methoxy-, methyl ester, acmc-209igx, methyl 3-bromo-p-anisate, rarechem al bf 0247, methyl-3-bromo-4-methoxybenzoate, 3-bromo-p-anisic acid methyl ester, #, methyl 3-bromo-4-methyloxy benzoate
PubChem CID 611662
IUPAC Name methyl 3-bromo-4-methoxybenzoate
SMILES COC(=O)C1=CC(Br)=C(OC)C=C1

Specifications

Melting Point 95°C to 98°C
Quantity 10 g
Formula Weight 245.08
Percent Purity 98%
Chemical Name or Material Methyl 3-bromo-4-methoxybenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only