missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Bromo-2-(bromomethyl)propionate 97.0+%, TCI America™

Catalog No. M20665G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20665G 5 g
M206625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20665G Supplier TCI America Supplier No. M20665G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22262-60-8
Molecular Formula C5H8Br2O2
Molecular Weight (g/mol) 259.925
MDL Number MFCD00011534
InChI Key USXVPPOARMSYGY-UHFFFAOYSA-N
PubChem CID 140895
IUPAC Name methyl 3-bromo-2-(bromomethyl)propanoate
SMILES COC(=O)C(CBr)CBr

Specifications

Boiling Point 62°C
Quantity 5 g
Synonym methyl 3-bromo-2-bromomethyl propionate, methyl 3-bromo-2-bromomethyl propanoate, methyl3-bromo-2-bromomethyl propionate, methyl bis bromomethyl acetate, methyl 2-bis-bromomethyl-acetate, propanoic acid, 3-bromo-2-bromomethyl-, methyl ester, propanoic acid,3-bromo-2-bromomethyl-, methyl ester, zlchem 71, acmc-1chw2, 3-bromo-2-bromomethyl-propionic acid methyl ester
Formula Weight 259.93
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Bromo-2-(bromomethyl)propionate