missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Amino-4-methylthiophene-2-carboxylate 98.0+%, TCI America™

Catalog No. M17725G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17725G 5 g
M177225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17725G Supplier TCI America Supplier No. M17725G
Only null left

Chemical Identifiers

CAS 85006-31-1
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.21
MDL Number MFCD00051822
InChI Key YICRPERKKBDRSP-UHFFFAOYSA-N
Synonym methyl3-amino-4-methylthiophene-2-carboxylate, 3-amino-4-methylthiophene-2-carboxylic acid methyl ester, unii-3la24473as, 3-amino-4-methyl-2-thiophenecarboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-amino-4-methyl-, methyl ester, methyl 3-amino-4-methyl-2-thenoate, methyl 3-amino-4-methyl-2-thiophenecarboxylate, methyl-3-amino-4-methylthiophene-2-carboxylate, methyl 3-amino-4-methyl-thiophene-2-carboxylate, 3-amino-4-methyl-thiophene-2-carboxylic acid methyl ester
PubChem CID 123584
IUPAC Name methyl 3-amino-4-methylthiophene-2-carboxylate
SMILES COC(=O)C1=C(N)C(C)=CS1

Specifications

Melting Point 86°C
Color White-Yellow
Quantity 5 g
Formula Weight 171.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Amino-4-methylthiophene-2-carboxylate