missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Amino-4-chlorobenzoate 98.0+%, TCI America™

Catalog No. M17785G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17785G 5 g
M177825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17785G Supplier TCI America Supplier No. M17785G
Only null left

Chemical Identifiers

CAS 40872-87-5
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.607
InChI Key LOCJPOYKBUUVKU-UHFFFAOYSA-N
Synonym 3-Amino-4-chlorobenzoic Acid Methyl Ester
PubChem CID 593055
IUPAC Name methyl 3-amino-4-chlorobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)Cl)N

Specifications

Melting Point 83°C
Color Yellow
Quantity 5 g
Formula Weight 185.61
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Amino-4-chlorobenzoate
EINECSNumber (3)-1481
TSCA No