missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-Dibromobenzoate 98.0+%, TCI America™

Catalog No. M19295G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19295G 5 g
M192925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19295G Supplier TCI America Supplier No. M19295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51329-15-8
Molecular Formula C8H6Br2O2
Molecular Weight (g/mol) 293.94
MDL Number MFCD00082648
InChI Key GSMAWUZTAIOCPL-UHFFFAOYSA-N
Synonym methyl3,5-dibromobenzoate, 3,5-dibromobenzoic acid methyl ester, benzoic acid, 3,5-dibromo-, methyl ester, 3,5-dibromomethylbenzoate, 1,3-dibromo-5-methoxycarbonyl benzene, pubchem3973, maybridge1_007706, acmc-209kt2, ksc274i7j, rarechem al bf 0770
PubChem CID 621923
IUPAC Name methyl 3,5-dibromobenzoate
SMILES COC(=O)C1=CC(Br)=CC(Br)=C1

Specifications

Melting Point 63°C
Quantity 5 g
Formula Weight 293.94
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3,5-Dibromobenzoate