missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-Dibromo-4-hydroxybenzoate Monohydrate 99.0+%, TCI America™

Catalog No. D250125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D250125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D250125G Supplier TCI America Supplier No. D250125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41727-47-3
Molecular Formula C8H6Br2O3
Molecular Weight (g/mol) 309.941
MDL Number MFCD00016416
InChI Key NVGJGYZKXBLIKY-UHFFFAOYSA-N
Synonym 3,5-dibromo-4-hydroxybenzoic acid methyl ester, 3,5-dibromo-4-hydroxy-benzoic acid methyl ester, methyl 3,5-bis bromanyl-4-oxidanyl-benzoate, acmc-20akhs, pubchem3978, methyl dibromosalicylate, rarechem al bf 0231, timtec-bb sbb012530, labotest-bb lt00455670, labotest-bb lt03332530
PubChem CID 726975
IUPAC Name methyl 3,5-dibromo-4-hydroxybenzoate
SMILES COC(=O)C1=CC(=C(C(=C1)Br)O)Br

Specifications

Melting Point 124°C
Color White
Quantity 25 g
Formula Weight 309.94
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3,5-Dibromo-4-hydroxybenzoate Monohydrate