missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-diaminobenzoate, 99%

Catalog No. AAL1084818
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1084818 50 g
AAL1084809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1084818 Supplier Thermo Scientific Chemicals Supplier No. L1084818
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1949-55-9 | C8H10N2O2 | 166.18 g/mol

Methyl 3,5-diaminobenzoate is used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3,5-diaminobenzoate is used as a primary and secondary intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong acids and strong oxidizing agents.

Chemical Identifiers

CAS 1949-55-9
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00041433
InChI Key IYWLDOMAEQHKJZ-UHFFFAOYSA-N
Synonym methyl 3,5-diaminobenzenecarboxylate, 3,5-diaminobenzoic acid methyl ester, methyldiaminobenzenecarboxylate, 3,5-diamino-benzoic acid methyl ester, benzoic acid,3,5-diamino-, methyl ester
PubChem CID 7010431
IUPAC Name methyl 3,5-diaminobenzoate
SMILES COC(=O)C1=CC(=CC(=C1)N)N

Specifications

Melting Point 131°C to 134°C
Odor Odorless
Quantity 50 g
Beilstein 2363309
Solubility Information Insoluble in water.
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material Methyl 3,5-diaminobenzoate
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only