missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-diamino-6-chloropyrazine-2-carboxylate, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022965
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022965 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022965 Supplier Merck Emd Millipore Supplier No. BP920

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

CAS 1458-01-1
Melting Point 211°C to 215°C (literature)
Molecular Formula C6H7ClN4O2
Linear Formula C6H7ClN4O2
MDL Number MFCD01928388
Quantity 10 mg
Formula Weight 202.60
Grade BP