missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4-Dihydroxybenzoate 98.0+%, TCI America™

Catalog No. M19435G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19435G 5 g
M194325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19435G Supplier TCI America Supplier No. M19435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2150-43-8
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00016437
InChI Key CUFLZUDASVUNOE-UHFFFAOYSA-N
Synonym methyl protocatechuate, protocatechuic acid, methyl ester, 3,4-dihydroxybenzoic acid methyl ester, protocatechuic acid methyl ester, benzoic acid, 3,4-dihydroxy-, methyl ester, 3,4-dihydroxymethylbenzoate, 3,4-dihydroxybenzoic acid methyl, mdhb, methyl 3,4-dihydroxy, acmc-1cfbz
PubChem CID 287064
IUPAC Name methyl 3,4-dihydroxybenzoate
SMILES COC(=O)C1=CC=C(O)C(O)=C1

Specifications

Melting Point 136°C
Color White
Quantity 5 g
Formula Weight 168.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3,4-Dihydroxybenzoate