missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4-diaminobenzoate, 98%

Catalog No. p-7022059
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1465606 5 g
AAA1465614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1465606 Supplier Thermo Scientific Chemicals Supplier No. A1465606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 36692-49-6 | C8H10N2O2 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36692-49-6
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00017098
InChI Key IOPLHGOSNCJOOO-UHFFFAOYSA-N
Synonym 3,4-diaminobenzoic acid methyl ester, methyl3,4-diaminobenzoate, methyl-3,4-diaminobenzoate, benzoic acid, 3,4-diamino-, methyl ester, 3,4-diamino-benzoic acid methyl ester, methyl 3,4-diaminobenzenecarboxylate, pubchem4610, acmc-1bn7s, methyl 3,4-diamino-benzoate, methyl-3,4-diamino-benzoate
PubChem CID 135524
IUPAC Name methyl 3,4-diaminobenzoate
SMILES COC(=O)C1=CC=C(N)C(N)=C1

Specifications

Melting Point 105°C to 108°C
Quantity 5 g
Beilstein 2091675
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material Methyl 3,4-diaminobenzoate
Hazard Category H301-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280i-P301+P310-P305+P351+P338-P330-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only