missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(4-Bromophenyl)propionate 98.0+%, TCI America™

Catalog No. M25885G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25885G 5 g
M25881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25885G Supplier TCI America Supplier No. M25885G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75567-84-9
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.1
MDL Number MFCD09953150
InChI Key FKPYNBFWCSTPOT-UHFFFAOYSA-N
PubChem CID 11447883
IUPAC Name methyl 3-(4-bromophenyl)propanoate
SMILES COC(=O)CCC1=CC=C(C=C1)Br

Specifications

Color Yellow
Boiling Point 92°C
Quantity 5 g
Synonym methyl 3-4-bromophenyl propanoate, methyl 3-4-bromophenyl propionate, 3-4-bromophenyl propanoic acid methyl ester, benzenepropanoic acid, 4-bromo-, methyl ester, 3-4-bromophenyl propionic acid methyl ester, acmc-209oyw, 3-4-bromophenyl propionate, ksc498a6l, methyl3-4-bromophenyl propionate, 3-4-bromophenyl-propionic acid methyl ester
Formula Weight 243.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-(4-Bromophenyl)propionate