missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3,4,5-Trimethoxybenzoate 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 82°C |
| Color | White |
| Boiling Point | 185°C |
| Quantity | 500 g |
| Formula Weight | 226.23 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Methyl 3,4,5-Trimethoxybenzoate |
Safety and Handling
| RTECSNumber | DI0820000 |
| TSCA | Yes |