missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4,5-trimethoxybenzoate, 98+%

Catalog No. AAA1193336
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1193336 500 g
AAA1193314 25 g
AAA1193322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1193336 Supplier Thermo Scientific Chemicals Supplier No. A1193336
Only null left

CAS: 1916-07-0 | C11H14O5 | 226.228 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1916-07-0
Molecular Formula C11H14O5
Molecular Weight (g/mol) 226.228
MDL Number MFCD00008431
InChI Key KACHFMOHOPLTNX-UHFFFAOYSA-N
Synonym trimethylgallic acid methyl ester, methyl tri-o-methylgallate, benzoic acid, 3,4,5-trimethoxy-, methyl ester, 3,4,5-trimethoxybenzoic acid methyl ester, unii-qfp1lsc7tk, 3,4,5-trimethoxybenzoic acid, methyl ester, qfp1lsc7tk, methyl3,4,5-trimethoxybenzoate, methyl 3,4,5-trimethoxy benzoate, acmc-209ev8
PubChem CID 15956
IUPAC Name methyl 3,4,5-trimethoxybenzoate
SMILES COC1=CC(=CC(=C1OC)OC)C(=O)OC

Specifications

Melting Point 81°C to 85°C
Boiling Point 274°C to 275°C
Quantity 500 g
Beilstein 2218156
Formula Weight 226.23
Percent Purity ≥98%
Chemical Name or Material Methyl 3,4,5-trimethoxybenzoate
EINECSNumber 217-629-3
RTECSNumber DI0820000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only