missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3,4,5-trihydroxybenzoate, 98%
Description
Methyl 3,4,5-trihydroxybenzoate exhibits potent antitumor activities by inhibiting tumor infiltration of CD4+CD25+ regulatory T cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 3,4,5-trihydroxybenzoate exhibits potent antitumor activities by inhibiting tumor infiltration of CD4+CD25+ regulatory T cells.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
Methyl 3,4,5-trihydroxybenzoate exhibits potent antitumor activities by inhibiting tumor infiltration of CD4+CD25+ regulatory T cells.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 198°C to 203°C |
| Quantity | 250 g |
| Beilstein | 2113180 |
| Merck Index | 144345 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 184.15 |
| Percent Purity | 0.98 |
| Chemical Name or Material | Methyl 3,4,5-trihydroxybenzoate |
Safety and Handling
| missing translation for 'healthHazard1' | H302-H315-H317-H319 |
| missing translation for 'healthHazard2' | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| missing translation for 'healthHazard3' | P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c |
| EINECSNumber | 202-741-7 |
| RTECSNumber | LW8000000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only