missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-[3-(dimethylamino)propoxy]benzoate, 97%, Thermo Scientific™

Numéro de catalogue. CC59422DE
Modifier l'affichage
Click to view available options
Quantité:
1g
5g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC59422DE 5g Amber Glass Bottle
CC59422DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC59422DE Fournisseur Thermo Scientific Chemicals Code fournisseur CC59422DE

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 921938-75-2
Formule moléculaire C13H19NO3
Poids moléculaire (g/mol) 237.299
Numéro MDL MFCD09702393
Clé InChI WGAZXKVAWOAUOI-UHFFFAOYSA-N
Synonyme methyl 3-3-dimethylamino propoxy benzoate
PubChem CID 24229605
Nom de l’IUPAC methyl 3-[3-(dimethylamino)propoxy]benzoate
SOURIRES CN(C)CCCOC1=CC=CC(=C1)C(=O)OC

Spécifications

Nom ou substance chimique Methyl 3-[3-(dimethylamino)propoxy]benzoate
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 5g
Poids de formule 237.3
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.