missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,3-dimethoxypropionate, 99%, Thermo Scientific™

Catalog No. AC264340050
Change view
Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC264340050 5g Glass bottle
AC264340250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC264340050 Supplier Thermo Scientific Chemicals Supplier No. 264340050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 7424-91-1
Molecular Formula C6H12O4
Molecular Weight (g/mol) 148.16
MDL Number MFCD00010650
InChI Key SMCVPMKCDDNUCQ-UHFFFAOYSA-N
Synonym methyl 3,3-dimethoxypropionate, 3,3-dimethoxypropionic acid methyl ester, methyl3,3-dimethoxypropionate, propanoic acid, 3,3-dimethoxy-, methyl ester, zlchem 282, pubchem20228, acmc-1bdjk, ksc490c4j, methyl 3,3-dimethoxy-propanoate, smcvpmkcddnucq-uhfffaoysa
PubChem CID 81924
IUPAC Name methyl 3,3-dimethoxypropanoate
SMILES COC(CC(=O)OC)OC

Specifications

Density 1.0600g/mL
Boiling Point 166°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4090 to 1.411
Linear Formula (CH3O)2CHCH2CO2CH3
Quantity 5g
Beilstein 03,1171
Specific Gravity 1.06
Viscosity 2.5 mPa.s (20°C)
Formula Weight 148.16
Percent Purity 99%
Chemical Name or Material Methyl 3, 3-dimethoxypropionate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 231-055-

RUO – Research Use Only