missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(trifluoromethyl)pyrimidine-4-carboxylate, 97%, Thermo Scientific Chemicals

Catalog No. p-7035110
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5281803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5281803 Supplier Thermo Scientific Chemicals Supplier No. H5281803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 878745-51-8 | Molecular Formula: C7H5F3N2O2 | Molecular Weight: 206.12 g/mol

Methyl 2-(trifluoromethyl)pyrimidine-4-carboxylate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-(trifluoromethyl)pyrimidine-4-carboxylate is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 878745-51-8
Molecular Formula C7H5F3N2O2
Molecular Weight (g/mol) 206.12
MDL Number MFCD05662690
InChI Key VBFLWQBZJODKRL-UHFFFAOYSA-N
Synonym methyl 2-trifluoromethyl pyrimidine-4-carboxylate, 2-trifluoromethyl-pyrimidine-4-carboxylic acid methyl ester, methyl2-trifluoromethyl pyrimidine-4-carboxylate, methyl 2-trifluoromethyl-4-pyrimidine carboxylate, 4-pyrimidinecarboxylic acid, 2-trifluoromethyl-, methyl ester, 4-pyrimidinecarboxylicacid, 2-trifluoromethyl-, methyl ester, 4-pyrimidinecarboxylicacid,2-trifluoromethyl-,methyl ester
PubChem CID 17750127
IUPAC Name methyl 2-(trifluoromethyl)pyrimidine-4-carboxylate
SMILES COC(=O)C1=NC(=NC=C1)C(F)(F)F

Specifications

Quantity 1 g
Formula Weight 206.12
Percent Purity 97%
Chemical Name or Material Methyl 2-(trifluoromethyl)pyrimidine-4-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only