missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Oxovalerate 90.0+%, TCI America™

Catalog No. M18635G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18635G 5 g
M186325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18635G Supplier TCI America Supplier No. M18635G
Only null left

Chemical Identifiers

CAS 6376-59-6
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
InChI Key MBNLVYPIKSWXCT-UHFFFAOYSA-N
Synonym 2-Oxovaleric Acid Methyl Ester, Methyl 2-Oxopentanoate, 2-Oxopentanoic Acid Methyl Ester
PubChem CID 642256
IUPAC Name methyl 2-oxopentanoate
SMILES CCCC(=O)C(=O)OC

Specifications

Color Yellow
Boiling Point 62°C
Quantity 5 g
Formula Weight 130.14
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Oxovalerate
TSCA No
Recommended Storage Refrigerator