missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-methylpropionimidate hydrochloride, 97%

Catalog No. AAH63386MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63386MD 250 mg
AAH6338603 1 g
AAH6338606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63386MD Supplier Thermo Scientific Chemicals Supplier No. H63386MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 39739-60-1 | C5H12ClNO | 137.61 g/mol

Methyl 2-methylpropionimidate hydrochloride, is used as a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-methylpropionimidate hydrochloride, is used as a fine chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 39739-60-1
Molecular Formula C5H12ClNO
Molecular Weight (g/mol) 137.61
MDL Number MFCD03844704
InChI Key RMXGPHAHPIIESQ-UHFFFAOYSA-N
Synonym methyl 2-methylpropanecarboximidate hydrochloride, methyl 2-methylpropanimidate hydrochloride, methyl 2-methylpropanimidic acid hydrochloride, methyl 2-methylpropanimidoate hydrochloride, methyl isobutyrimidate hydrochloride, methyl 2-methylpropanimidate-hydrogen chloride 1/1, methyl 1z-2-methylpropanimidoate hydrochloride 1:1
PubChem CID 371709
IUPAC Name methyl 2-methylpropanimidate;hydrochloride
SMILES Cl.COC(=N)C(C)C

Specifications

Color White
Linear Formula (CH3)2CHC(=NH)OCH3·HCl
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 137.61
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 2-methylpropionimidate hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only