missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-methyl-1,3-benzoxazole-5-carboxylate, 97%

Catalog No. AC432400050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432400050 5 g Glass bottle
AC432400010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432400050 Supplier Thermo Scientific Chemicals Supplier No. 432400050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 136663-21-3 | C10H9NO3 | 191.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 136663-21-3
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.19
MDL Number MFCD07774245
InChI Key UFLDNRAXHGPDOD-UHFFFAOYSA-N
Synonym methyl 2-methylbenzo d oxazole-5-carboxylate, 5-benzoxazolecarboxylicacid, 2-methyl-, methyl ester, acmc-1c498, methyl2-methyl-1,3-benzoxazole-5-carboxylate, methyl 2-methylbenzoxazole-5-carboxylate, 5-methoxycarbonyl-2-methyl-1,3-benzoxazole, methyl 2-methyl-1?3-benzoxazole-5-carboxylate, 2-methylbenzoxazole-5-carboxylic acid methyl ester, 5-benzoxazolecarboxylicacid,2-methyl-,methyl ester, 5-benzoxazolecarboxylic acid, 2-methyl-, methyl ester
PubChem CID 12157466
IUPAC Name methyl 2-methyl-1,3-benzoxazole-5-carboxylate
SMILES CC1=NC2=C(O1)C=CC(=C2)C(=O)OC

Specifications

Melting Point 67°C to 68°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 191.19
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-methyl-1,3-benzoxazole-5-carboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

RUO – Research Use Only