missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-chloroacetoacetate, 95%, Thermo Scientific™

Numéro de catalogue. p-2691715
Modifier l'affichage
Click to view available options
Quantité:
10mL
50mL
Emballage:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC179900100 10mL Glass bottle
AC179900500 50mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC179900100 Fournisseur Thermo Scientific Chemicals Code fournisseur 179900100

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 4755-81-1
Formule moléculaire C5H7ClO3
Poids moléculaire (g/mol) 150.56
Numéro MDL MFCD00008757
Clé InChI GYQRIAVRKLRQKP-UHFFFAOYSA-N
Synonyme methyl 2-chloroacetoacetate, butanoic acid, 2-chloro-3-oxo-, methyl ester, 2-chloroacetoacetic acid methyl ester, 2-chloro-3-oxo-butyric acid methyl ester, methyl2-chloroacetoacetate, acmc-209k9v, methyl-2-chloroacetoacetate, methyl-alpha-chloroacetoacetate, 2-chloroacetoacetate methyl ester, gyqriavrklrqkp-uhfffaoysa
PubChem CID 107332
Nom de l’IUPAC methyl 2-chloro-3-oxobutanoate
SOURIRES CC(=O)C(C(=O)OC)Cl

Spécifications

Nom ou substance chimique Methyl 2-chloroacetoacetate
Densité 1.2400g/mL
Point d’ébullition 137°C
Point d’éclair 72°C
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 95%
Emballage Glass bottle
Indice de réfraction 1.4455 to 1.4475
Formule linéaire CH3COCH(Cl)CO2CH3
Quantité 10mL
Gravité spécifique 1.24
Renseignements sur la solubilité Solubility in water: 50g/L (20°C). Other solubilities: soluble in organic solvents
Poids de formule 150.56
Pourcentage de pureté 95%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-286-6

RUO – Research Use Only