missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Chloro-5-nitrobenzoate 98.0+%, TCI America™
$42.36 - $132.84
Chemical Identifiers
| CAS | 6307-82-0 |
|---|---|
| Molecular Formula | C8H6ClNO4 |
| Molecular Weight (g/mol) | 215.589 |
| MDL Number | MFCD00184587 |
| InChI Key | VCYWZLGOWNCJNJ-UHFFFAOYSA-N |
| Synonym | 2-chloro-5-nitrobenzoic acid methyl ester, benzoic acid, 2-chloro-5-nitro-, methyl ester, methyl 2-chloro-5-nitrobenzenecarboxylate, 2-chloro-5-nitro-benzoic acid methyl ester, methylester kyseliny 2-chlor-5-nitrobenzoove czech, pubchem3711, acmc-209ncv, methylester kyseliny 2-chlor-5-nitrobenzoove, 4-09-00-01228 beilstein handbook reference, methyl2-chloro-5-nitrobenzoate |
| PubChem CID | 22754 |
| IUPAC Name | methyl 2-chloro-5-nitrobenzoate |
| SMILES | COC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])Cl |
Chemical Identifiers
| 6307-82-0 | |
| 215.589 | |
| VCYWZLGOWNCJNJ-UHFFFAOYSA-N | |
| 22754 | |
| COC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])Cl |
| C8H6ClNO4 | |
| MFCD00184587 | |
| 2-chloro-5-nitrobenzoic acid methyl ester, benzoic acid, 2-chloro-5-nitro-, methyl ester, methyl 2-chloro-5-nitrobenzenecarboxylate, 2-chloro-5-nitro-benzoic acid methyl ester, methylester kyseliny 2-chlor-5-nitrobenzoove czech, pubchem3711, acmc-209ncv, methylester kyseliny 2-chlor-5-nitrobenzoove, 4-09-00-01228 beilstein handbook reference, methyl2-chloro-5-nitrobenzoate | |
| methyl 2-chloro-5-nitrobenzoate |
Specifications
| 6307-82-0 | |
| C8H6ClNO4 | |
| 5 g | |
| VCYWZLGOWNCJNJ-UHFFFAOYSA-N | |
| methyl 2-chloro-5-nitrobenzoate | |
| 22754 | |
| ≥98.0% (GC) | |
| Methyl 2-Chloro-5-nitrobenzoate |
| 70°C | |
| MFCD00184587 | |
| 2-chloro-5-nitrobenzoic acid methyl ester, benzoic acid, 2-chloro-5-nitro-, methyl ester, methyl 2-chloro-5-nitrobenzenecarboxylate, 2-chloro-5-nitro-benzoic acid methyl ester, methylester kyseliny 2-chlor-5-nitrobenzoove czech, pubchem3711, acmc-209ncv, methylester kyseliny 2-chlor-5-nitrobenzoove, 4-09-00-01228 beilstein handbook reference, methyl2-chloro-5-nitrobenzoate | |
| COC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])Cl | |
| 215.589 | |
| 215.59 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : DG5426500
TSCA : No