CAS RN 6307-82-0
IUPAC Name:
methyl 2-chloro-5-nitrobenzoate
Synonyms:
methyl 2-chloro-5-nitrobenzoate
Molecular Weight (g/mol):
215.59
Molecular Formula:
C8H6ClNO4
InChi Key:
VCYWZLGOWNCJNJ-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(=CC=C1Cl)[N+]([O-])=O
1
–
1
of
1
results
1
Methyl 2-Chloro-5-nitrobenzoate 98.0+%, TCI America™
CAS: 6307-82-0 Molecular Formula: C8H6ClNO4 Molecular Weight (g/mol): 215.589 MDL Number: MFCD00184587 InChI Key: VCYWZLGOWNCJNJ-UHFFFAOYSA-N Synonym: 2-chloro-5-nitrobenzoic acid methyl ester,benzoic acid, 2-chloro-5-nitro-, methyl ester,methyl 2-chloro-5-nitrobenzenecarboxylate,2-chloro-5-nitro-benzoic acid methyl ester,methylester kyseliny 2-chlor-5-nitrobenzoove czech,pubchem3711,acmc-209ncv,methylester kyseliny 2-chlor-5-nitrobenzoove,4-09-00-01228 beilstein handbook reference,methyl2-chloro-5-nitrobenzoate PubChem CID: 22754 IUPAC Name: methyl 2-chloro-5-nitrobenzoate SMILES: COC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])Cl