missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Chloro-5-nitrobenzoate 98.0+%, TCI America™

Catalog No. c173225g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
C173225G 25 g
C17325G 5 g
2 options

Catalog No. C173225G

Supplier: TCI America C173225G

Only null left

Chemical Identifiers

CAS 6307-82-0
Molecular Formula C8H6ClNO4
Molecular Weight (g/mol) 215.589
MDL Number MFCD00184587
InChI Key VCYWZLGOWNCJNJ-UHFFFAOYSA-N
Synonym 2-chloro-5-nitrobenzoic acid methyl ester, benzoic acid, 2-chloro-5-nitro-, methyl ester, methyl 2-chloro-5-nitrobenzenecarboxylate, 2-chloro-5-nitro-benzoic acid methyl ester, methylester kyseliny 2-chlor-5-nitrobenzoove czech, pubchem3711, acmc-209ncv, methylester kyseliny 2-chlor-5-nitrobenzoove, 4-09-00-01228 beilstein handbook reference, methyl2-chloro-5-nitrobenzoate
PubChem CID 22754
IUPAC Name methyl 2-chloro-5-nitrobenzoate
SMILES COC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])Cl

Specifications

Melting Point 70°C
Quantity 25 g
Formula Weight 215.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Chloro-5-nitrobenzoate
RTECSNumber DG5426500
TSCA No