missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Bromo-5-nitrobenzoate 98.0+%, TCI America™

Catalog No. B19165G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19165G 5 g
B191625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19165G Supplier TCI America Supplier No. B19165G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6942-36-5
Molecular Formula C8H6BrNO4
Molecular Weight (g/mol) 260.043
MDL Number MFCD00010867
InChI Key VSEYYEKRZNRECT-UHFFFAOYSA-N
Synonym 2-bromo-5-nitrobenzoic acid methyl ester, benzoic acid, 2-bromo-5-nitro-, methyl ester, methyl 2-bromo-5-nitro-benzoate, 2-bromo-5-nitro-benzoic acid methyl ester, methyl2-bromo-5-nitrobenzoate, pubchem3971, acmc-1b9le, ksc495k5r, methyl 5-nitro-2-bromobenzoate, methyl-2-bromo-5-nitrobenzoate
PubChem CID 245494
IUPAC Name methyl 2-bromo-5-nitrobenzoate
SMILES COC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])Br

Specifications

Melting Point 82°C
Color Yellow
Quantity 5 g
Formula Weight 260.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Bromo-5-nitrobenzoate