missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Bromo-5-chlorobenzoate 98.0+%, TCI America™

Catalog No. M20335G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20335G 5 g
M203325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20335G Supplier TCI America Supplier No. M20335G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 27007-53-0
Molecular Formula C8H6BrClO2
Molecular Weight (g/mol) 249.488
MDL Number MFCD00144763
InChI Key BIECSXCXIXHDBC-UHFFFAOYSA-N
Synonym 2-bromo-5-chlorobenzoic acid methyl ester, benzoic acid, 2-bromo-5-chloro-, methyl ester, pubchem11238, acmc-1cr8r, ksc494q3f, methyl-2-bromo-5-chlorobenzoate, methyl 2-bromo-5-chloro-benzoate, methyl 2-bromanyl-5-chloranyl-benzoate, 2-bomo-5-chlorobenzoic acid methyl ester
PubChem CID 280500
IUPAC Name methyl 2-bromo-5-chlorobenzoate
SMILES COC(=O)C1=C(C=CC(=C1)Cl)Br

Specifications

Melting Point 41°C
Color White-Yellow
Quantity 5 g
Formula Weight 249.49
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material Methyl 2-Bromo-5-chlorobenzoate