missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-bromo-5-chlorobenzoate, 98%

Catalog No. AAB2530922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2530922 100 g
AAB2530914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2530922 Supplier Thermo Scientific Chemicals Supplier No. B2530922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 27007-53-0 | C8H6BrClO2 | 249.488 g/mol

Methyl 2-bromo-5-chlorobenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-bromo-5-chlorobenzoate is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 27007-53-0
Molecular Formula C8H6BrClO2
Molecular Weight (g/mol) 249.488
MDL Number MFCD00144763
InChI Key BIECSXCXIXHDBC-UHFFFAOYSA-N
Synonym 2-bromo-5-chlorobenzoic acid methyl ester, benzoic acid, 2-bromo-5-chloro-, methyl ester, pubchem11238, acmc-1cr8r, ksc494q3f, methyl-2-bromo-5-chlorobenzoate, methyl 2-bromo-5-chloro-benzoate, methyl 2-bromanyl-5-chloranyl-benzoate, 2-bomo-5-chlorobenzoic acid methyl ester
PubChem CID 280500
IUPAC Name methyl 2-bromo-5-chlorobenzoate
SMILES COC(=O)C1=C(C=CC(=C1)Cl)Br

Specifications

Melting Point 37°C to 40°C
Quantity 100 g
Solubility Information Slightly soluble in water.
Formula Weight 249.49
Percent Purity 98%
Chemical Name or Material Methyl 2-bromo-5-chlorobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only