missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Aminothiazole-5-carboxylate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 191°C |
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 158.18 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Methyl 2-Aminothiazole-5-carboxylate |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |