missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-5-(trifluoromethyl)benzoate, 98%

Catalog No. AAH6428303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6428303 1 g
AAH6428306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6428303 Supplier Thermo Scientific Chemicals Supplier No. H6428303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 117324-58-0 | C9H8F3NO2 | 219.163 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 117324-58-0
Molecular Formula C9H8F3NO2
Molecular Weight (g/mol) 219.163
MDL Number MFCD08234902
InChI Key QGFUDNJZHZNPCS-UHFFFAOYSA-N
PubChem CID 14233808
IUPAC Name methyl 2-amino-5-(trifluoromethyl)benzoate
SMILES COC(=O)C1=C(C=CC(=C1)C(F)(F)F)N

Specifications

Color White
Quantity 1 g
Synonym methyl 2-amino-5-trifluoromethyl benzoate, methyl2-amino-5-trifluoromethyl benzoate, 2-amino-5-trifluoromethyl benzoic acid methyl ester, benzoic acid,2-amino-5-trifluoromethyl-, methyl ester, benzoic acid, 2-amino-5-trifluoromethyl-, methyl ester, acmc-209z2m, benzoicacid,2-amino-5-trifluoromethyl-,methylester, methyl 5-trifluoromethylanthranilate, 2-amino-5-trifluoromethyl-benzoic acid methyl ester
Formula Weight 219.16
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 2-amino-5-(trifluoromethyl)benzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only