missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Amino-3-methylbenzoate 98.0+%, TCI America™
$86.64 - $338.84
Chemical Identifiers
| CAS | 22223-49-0 |
|---|---|
| Molecular Formula | C9H11NO2 |
| Molecular Weight (g/mol) | 165.192 |
| MDL Number | MFCD06200918 |
| InChI Key | VSFYTPXXMLJNAU-UHFFFAOYSA-N |
| Synonym | methyl 3-methylanthranilate, 2-amino-3-methylbenzoic acid methyl ester, benzoic acid, 2-amino-3-methyl-, methyl ester, methyl 2-amino-3-methylbenzenecarboxylate, 2-amino-3-methyl-benzoic acid methyl ester, methyl2-amino-3-methylbenzoate, pubchem10929, acmc-209ftn, methyl 2-amino-m-toluate, ksc495o6t |
| PubChem CID | 2763406 |
| IUPAC Name | methyl 2-amino-3-methylbenzoate |
| SMILES | CC1=CC=CC(=C1N)C(=O)OC |
Chemical Identifiers
| 22223-49-0 | |
| 165.192 | |
| VSFYTPXXMLJNAU-UHFFFAOYSA-N | |
| 2763406 | |
| CC1=CC=CC(=C1N)C(=O)OC |
| C9H11NO2 | |
| MFCD06200918 | |
| methyl 3-methylanthranilate, 2-amino-3-methylbenzoic acid methyl ester, benzoic acid, 2-amino-3-methyl-, methyl ester, methyl 2-amino-3-methylbenzenecarboxylate, 2-amino-3-methyl-benzoic acid methyl ester, methyl2-amino-3-methylbenzoate, pubchem10929, acmc-209ftn, methyl 2-amino-m-toluate, ksc495o6t | |
| methyl 2-amino-3-methylbenzoate |
Specifications
| 22223-49-0 | |
| 153°C | |
| MFCD06200918 | |
| methyl 3-methylanthranilate, 2-amino-3-methylbenzoic acid methyl ester, benzoic acid, 2-amino-3-methyl-, methyl ester, methyl 2-amino-3-methylbenzenecarboxylate, 2-amino-3-methyl-benzoic acid methyl ester, methyl2-amino-3-methylbenzoate, pubchem10929, acmc-209ftn, methyl 2-amino-m-toluate, ksc495o6t | |
| CC1=CC=CC(=C1N)C(=O)OC | |
| 165.192 | |
| 165.19 | |
| Crystalline Lumps |
| White-Yellow | |
| C9H11NO2 | |
| 5 g | |
| VSFYTPXXMLJNAU-UHFFFAOYSA-N | |
| methyl 2-amino-3-methylbenzoate | |
| 2763406 | |
| ≥98.0% (GC,T) | |
| Methyl 2-Amino-3-methylbenzoate |
Safety and Handling
TSCA : No