missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Amino-3-methylbenzoate 98.0+%, TCI America™
Supplier: TCI America M219325G
Specifications
| Methyl 2-Amino-3-methylbenzoate | |
| White-Yellow | |
| C9H11NO2 | |
| 25 g | |
| VSFYTPXXMLJNAU-UHFFFAOYSA-N | |
| methyl 2-amino-3-methylbenzoate | |
| 2763406 | |
| ≥98.0% (GC,T) |
| 22223-49-0 | |
| 153°C | |
| MFCD06200918 | |
| methyl 3-methylanthranilate, 2-amino-3-methylbenzoic acid methyl ester, benzoic acid, 2-amino-3-methyl-, methyl ester, methyl 2-amino-3-methylbenzenecarboxylate, 2-amino-3-methyl-benzoic acid methyl ester, methyl2-amino-3-methylbenzoate, pubchem10929, acmc-209ftn, methyl 2-amino-m-toluate, ksc495o6t | |
| CC1=CC=CC(=C1N)C(=O)OC | |
| 165.192 | |
| 165.19 | |
| Crystalline Lumps |
Chemical Identifiers
| 22223-49-0 | |
| 165.192 | |
| VSFYTPXXMLJNAU-UHFFFAOYSA-N | |
| 2763406 | |
| CC1=CC=CC(=C1N)C(=O)OC |
| C9H11NO2 | |
| MFCD06200918 | |
| methyl 3-methylanthranilate, 2-amino-3-methylbenzoic acid methyl ester, benzoic acid, 2-amino-3-methyl-, methyl ester, methyl 2-amino-3-methylbenzenecarboxylate, 2-amino-3-methyl-benzoic acid methyl ester, methyl2-amino-3-methylbenzoate, pubchem10929, acmc-209ftn, methyl 2-amino-m-toluate, ksc495o6t | |
| methyl 2-amino-3-methylbenzoate |
Safety and Handling
TSCA : No