missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,6-Dichlorophenylacetate 98.0+%, TCI America™

Catalog No. D23285G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23285G 5 g
D232825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23285G Supplier TCI America Supplier No. D23285G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 54551-83-6
Molecular Formula C9H8Cl2O2
Molecular Weight (g/mol) 219.061
MDL Number MFCD00191640
InChI Key FCWRUYPZZJPCCG-UHFFFAOYSA-N
PubChem CID 2734107
IUPAC Name methyl 2-(2,6-dichlorophenyl)acetate
SMILES COC(=O)CC1=C(C=CC=C1Cl)Cl

Specifications

Color Colorless
Quantity 5 g
Synonym methyl 2,6-dichlorophenylacetate, methyl 2-2,6-dichlorophenyl acetate, 2,6-dichlorophenylacetic acid methyl ester, benzeneacetic acid,2,6-dichloro-, methyl ester, benzeneacetic acid, 2,6-dichloro-, methyl ester, acmc-209lhf, methyl2,6-dichlorophenylacetate, guanfacine methyl ester impurity, methyl 2,6-dichlorophenyl acetate, 2,6-dichloro-phenylacetic acid methyl ester
Formula Weight 219.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2,6-Dichlorophenylacetate