missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,4-dihydroxybenzoate, 97%, Thermo Scientific™

Numéro de catalogue. p-3758388
Modifier l'affichage
Click to view available options
Quantité:
25g
100g
Emballage:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC126730250 25g Glass bottle
AC126731000 100g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC126730250 Fournisseur Thermo Scientific Chemicals Code fournisseur 126730250

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 2150-47-2
Formule moléculaire C8H8O4
Poids moléculaire (g/mol) 168.15
Numéro MDL MFCD00002276
Clé InChI IIFCLXHRIYTHPV-UHFFFAOYSA-N
Synonyme 2,4-dihydroxybenzoic acid methyl ester, benzoic acid, 2,4-dihydroxy-, methyl ester, methyl beta-resorcylate, methyl-2,4-dihydroxybenzoate, unii-xe68bu3oyj, xe68bu3oyj, .beta.-resorcylic acid, methyl ester, beta-resorcylic acid methyl ester, methyl benzoate, 2,4-dihydroxy, methyl b-resorcylate
PubChem CID 16523
Nom de l’IUPAC methyl 2,4-dihydroxybenzoate
SOURIRES COC(=O)C1=C(C=C(C=C1)O)O

Spécifications

Nom ou substance chimique Methyl 2, 4-dihydroxybenzoate
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 97%
Emballage Glass bottle
Formule linéaire (HO)2C6H3CO2CH3
Quantité 25g
Beilstein 10,I,178
Poids de formule 168.15
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-428-3

RUO – Research Use Only