missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,3,4,6-Tetra-O-acetyl-1-thio-alpha-D-mannopyranoside (contains ca. 5% beta-isomer) 95.0+%, TCI America™

Catalog No. M15015G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M15015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M15015G Supplier TCI America Supplier No. M15015G

Chemical Identifiers

CAS 64550-71-6
Molecular Formula C15H22O9S
Molecular Weight (g/mol) 378.392
MDL Number MFCD00080808
InChI Key XWFUCHLBRWBKGN-NIFZNCRKSA-N
PubChem CID 11773340
IUPAC Name [(2R,3R,4S,5S,6R)-3,4,5-triacetyloxy-6-methylsulfanyloxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)SC)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Color White
Quantity 5 g
Formula Weight 378.39
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2,3,4,6-Tetra-O-acetyl-1-thio-alpha-D-mannopyranoside (contains ca. 5% beta-isomer)
TSCA No
Recommended Storage Freezer