missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1H-indazole-6-carboxylate, 95%, Thermo Scientific™

Catalog No. AAH3293303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3293303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3293303 Supplier Thermo Scientific Chemicals Supplier No. H3293303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Methyl 1H-indazole-6-carboxylate is used in organic synthesis.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.  

Chemical Identifiers

CAS 170487-40-8
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.175
MDL Number MFCD07371612
InChI Key TUSICEWIXLMXEY-UHFFFAOYSA-N
Synonym 1h-indazole-6-carboxylic acid methyl ester, methyl indazole-6-carboxylate, indazole-6-carboxylic acid methyl ester, 6-1h indazole carboxylic acid methyl ester, 1h-indazole-6-carboxylic acid, methyl ester, pubchem18064, methyl indazole-6carboxylate, ksc494o4p, methyl-1h-indazole-6-carboxylate, 2h-indazole-6-carboxylic acid methyl ester
PubChem CID 286535
IUPAC Name methyl 1H-indazole-6-carboxylate
SMILES COC(=O)C1=CC2=C(C=C1)C=NN2

Specifications

Melting Point 142°C to 144°C
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 176.17
Percent Purity 95%
Chemical Name or Material Methyl 1H-indazole-6-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures