missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-methylpyrrole-2-carboxylate, 99%

Catalog No. AAA1309514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1309514 25 g
AAA1309506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1309514 Supplier Thermo Scientific Chemicals Supplier No. A1309514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 37619-24-2 | C7H9NO2 | 139.154 g/mol

Methyl 1-methylpyrrole-2-carboxylate is used as chemical and organic Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 1-methylpyrrole-2-carboxylate is used as chemical and organic Intermediates.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 37619-24-2
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
MDL Number MFCD00052747
InChI Key APHVGKYWHWFAQV-UHFFFAOYSA-N
Synonym methyl 1-methyl-1h-pyrrole-2-carboxylate, methyl1-methyl-1h-pyrrole-2-carboxylate, 1-methyl-1h-pyrrole-2-carboxylic acid methyl ester, methyl 1-methyl-2-pyrrolecarboxylate, 1-methylpyrrole-2-carboxylic acid methyl ester, pubchem12438, acmc-209iue, methylmethylpyrrolecarboxylate, 2-methoxycarbonyl-1-methylpyrrole, methyl 1-methyl-pyrrole-2-carboxylate
PubChem CID 142178
IUPAC Name methyl 1-methylpyrrole-2-carboxylate
SMILES CN1C=CC=C1C(=O)OC

Specifications

Boiling Point 96°C to 98°C (15 mmHg)
Refractive Index 1.522
Quantity 25 g
Beilstein 116477
Solubility Information Slightly soluble in water.
Formula Weight 139.16
Percent Purity 99%
Chemical Name or Material Methyl 1-methylpyrrole-2-carboxylate
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only