missing translation for 'onlineSavingsMsg'
Learn More

Methoxyhydroquinone 98.0+%, TCI America™

Catalog No. M05395G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M05395G 5 g
M053925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M05395G Supplier TCI America Supplier No. M05395G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 824-46-4
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00013971
InChI Key LAQYHRQFABOIFD-UHFFFAOYSA-N
Synonym 2-methoxyhydroquinone, methoxyhydroquinone, 1,4-benzenediol, 2-methoxy, 1,4-dihydroxy-2-methoxybenzene, unii-2hi6hnr5u1, 2hi6hnr5u1, o-methoxyhydroquinone, 2,5-dihydroxyanisole, 2-mohydrop, acmc-209pp0
PubChem CID 69988
IUPAC Name 2-methoxybenzene-1,4-diol
SMILES COC1=C(C=CC(=C1)O)O

Specifications

Melting Point 89°C
Boiling Point 180°C
Quantity 5 g
Formula Weight 140.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methoxyhydroquinone