missing translation for 'onlineSavingsMsg'
Learn More

Methoxyacetyl chloride, 97%, stab. with ca 0.3% magnesium oxide

Catalog No. AAL0321014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0321014 25 g
AAL0321006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0321014 Supplier Thermo Scientific Chemicals Supplier No. L0321014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 38870-89-2 | C3H5ClO2 | 108.521 g/mol

Methoxyacetyl chloride is used as an intermediate for active pharmaceutical ingredients and dyes. Further, it acts as a precursor for the synthesis of agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methoxyacetyl chloride is used as an intermediate for active pharmaceutical ingredients and dyes. Further, it acts as a precursor for the synthesis of agrochemicals.

Solubility
Miscible with acetone, ether, carbon tetrachloride and chloroform.

Notes
Moisture sensitive. Store in a cool place. Incompatible with water, alcohols, oxidizing agents and strong bases.

Chemical Identifiers

CAS 38870-89-2
Molecular Formula C3H5ClO2
Molecular Weight (g/mol) 108.521
MDL Number MFCD00000728
InChI Key JJKWHOSQTYYFAE-UHFFFAOYSA-N
Synonym methoxyacetyl chloride, methoxyacetylchloride, acetyl chloride, methoxy, methoxy-acetyl chloride, methoxyacetic acid chloride, unii-t66s02b1s3, methoxy acetylchloride, 2-methoxyacetylchloride, methoxy acetyl chloride, ch3och2cocl
PubChem CID 96623
IUPAC Name 2-methoxyacetyl chloride
SMILES COCC(=O)Cl

Specifications

Melting Point <-40°C
Density 1.185
Boiling Point 112°C to 113°C
Flash Point 28°C (82°F)
Refractive Index 1.424
Quantity 25 g
UN Number UN2920
Beilstein 1740244
Sensitivity Moisture sensitive
Solubility Information Miscible with acetone,ether,carbon tetrachloride and chloroform.
Formula Weight 108.52
Percent Purity 97%
Chemical Name or Material Methoxyacetyl chloride, stabilized with ≈0.3% magnesium oxide
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H226
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 254-169-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only