missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Methoxy-X04
Description
Methoxy-X04 is a fluorescent amyloid β (Aβ) probe for the detection and quantification of plaques, tangles and cerebrovascular amyloid. Methoxy-X04 Displays high in vitro binding affinity (Ki = 26.8 nM); binds selectively to fibrillar β-sheet deposits. Methoxy-X04 is a derivative of Congo Red (Cat. No. 5167). Methoxy-X04 can be used for in vivo Aβ plaque labeling. Methoxy-X04 co-localizes with CD68+ phagosomes in plaque-associated Iba1+ microglia; and it labels molecularly distinct plaque-associated microglia populations. Brain penetrant.
Spécifications
Spécifications
| Quantity | 10 mg |
| Formula Weight | 344.4 |
| Purity | >98% |
| Molecular Weight (g/mol) | 344.41 |
| Chemical Name or Material | Methoxy-X04 |
Sécurité et manipulation
| Recommended Storage | Store at +4°C |