missing translation for 'onlineSavingsMsg'
Learn More

Methionol 99.0+%, TCI America™

Catalog No. M07355G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M07355G 5 g
M073525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M07355G Supplier TCI America Supplier No. M07355G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 505-10-2
Molecular Formula C4H10OS
Molecular Weight (g/mol) 106.18
MDL Number MFCD00036560
InChI Key CZUGFKJYCPYHHV-UHFFFAOYSA-N
Synonym methionol, 3-methylthiopropanol, 3-methylthio-1-propanol, 3-hydroxypropyl methyl sulfide, 3-methylthio propanol, 3-methylmercapto-1-propanol, 1-propanol, 3-methylthio, 3-methylsulfanyl propan-1-ol, gamma-methylmercaptopropyl alcohol
PubChem CID 10448
ChEBI CHEBI:49019
IUPAC Name 3-(methylsulfanyl)propan-1-ol
SMILES CSCCCO

Specifications

Color Yellow
Boiling Point 136°C
Quantity 5 g
Formula Weight 106.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methionol
EINECSNumber (2)-0469
TSCA Yes