missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methapyrilene HCl, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Packaging | Ampule |
| CAS | 135-23-9 |
| Physical Form | Neat |
| MDL Number | MFCD00050665 |
| Type | Pharmaceuticals, Illicit Drugs and Alcohol Standards |
| Quantity | 1000 mg |
| Grade | Analytical Standard |