missing translation for 'onlineSavingsMsg'
Learn More

Methanol, OmniSolv™, MilliporeSigma™

Catalog No. MMX04826
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MMX04826 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MMX04826 Supplier Technidata Supplier No. MX04826

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Purge and Trap Analysis

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -97.8°C
Color Colorless
Density 0.79g/cm3 at 20°C
Quantity 1 L
Flash Point Closed cup: 12°C (53.6°F), Open cup: 15.85°C (60.5°F)
Ignition Point 464°C (867.2°F)
Odor Alcohol-like
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 12.9 kPa (97mm Hg) [20°C]
UN Number 1230
Boiling Point 64.5°C
Solubility Information Soluble in water
Formula Weight 32.04
Percent Purity ≥99.9%
Chemical Name or Material Methanol
Show More Show Less
Recommended Storage Store in accordance with local regulations. Store in a segregated and approved area. Store in original container, protected from direct sunlight. Eliminate all ignition sources. Separate from oxidizing materials. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept uprightto prevent leakage.