missing translation for 'onlineSavingsMsg'
Learn More

Methanol, Histology grade, MilliporeSigma™

Catalog No. MMX04904
Change view
Click to view available options
Quantity:
4 L
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MMX04904 4 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MMX04904 Supplier Technidata Supplier No. MX04904

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Density 0.792 g/cc at 20°C
Quantity 4 L
Flash Point 10°C
Ignition Point 455°C (851°F)
Packaging Plastic Bottle
Physical Form Liquid
Vapor Pressure 128 hPa (20°C)
Water 0.5%
Boiling Point 64.5°C (1013 hPa)
Formula Weight 32.04
Percent Purity 98%
Grade Histological
Chemical Name or Material Methanol
Show More Show Less
Hazard Statement Hazard StatementsH225 Highly flammable liquid and vapor. H301 + H311 + H331 Toxic if swallowed, in contact with skin or if inhaled. H370 Causes damage to organs (Eyes).
Precautionary Statement Precautionary StatementsP210 Keep away from heat/sparks/open flames/hot surfaces. - No smoking. P233 Keep container tightly closed. P280 Wear protective gloves/ protective clothing. P302 + P352 IF ON SKIN: Wash with plenty of soap and water. P304 + P340 IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. P308 + P310 IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician. P403 + P235 Store in a well-ventilated place. Keep cool
EINECSNumber 200-659-6
RTECSNumber PC1400000
Recommended Storage Store at room temperature.