missing translation for 'onlineSavingsMsg'
Learn More

Methanol 99.8+%, TCI America™

Catalog No. M0628500ML
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0628500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M0628500ML Supplier TCI America Supplier No. M0628500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
IUPAC Name methanol

Specifications

Color Colorless
Quantity 500 mL
Physical Form Clear Liquid at 20°C
UN Number 1230
Boiling Point 64°C
Formula Weight 32.04
Percent Purity ≥99.8% (GC)
Chemical Name or Material Methanol [for HPLC Solvent]
EINECSNumber (2)-0201
RTECSNumber PC1400000
TSCA Yes