missing translation for 'onlineSavingsMsg'
Learn More

Methanol-d4, NMR Reference Standard, 99.8 atom % D, NMR Tube Size: 3 mm 8 in., MilliporeSigma™ Supelco™

Catalog No. 111017043
Change view
Click to view available options
Quantity:
0.235 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017043 0.235 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017043 Supplier Merck Emd Millipore Supplier No. 7219218IN

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging NMR Tube Size: 3 mm 8 in.
Linear Formula CD3OD
Quantity 0.235 mL
Formula Weight 36.07
Percent Purity 99% (CP)
Grade NMR Reference Standard