missing translation for 'onlineSavingsMsg'
Learn More

Methanol, LC-MS, for LC-MS and HPLC, >99.9%, Solstice

Catalog No. 6000962
Change view
Click to view available options
Quantity:
1 x 4 L
4 x 2.5 L
6 x 1 L
Packaging:
GL45 Glass Bottle
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000962 6 x 1 L GL45 Glass Bottle
6000960 4 x 2.5 L GL45 Glass Bottle
6000961 1 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6000962 Supplier Solstice Supplier No. LC2301

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

CAS Max % 100.0000%
Density 0.792 g/cm3
Quantity 6 x 1 L
Flash Point 11°C
Packaging GL45 Glass Bottle
UN Number UN1230
Water 0.05% max.
Beilstein 1098229
Boiling Point 64.7°C
Solubility Information Miscible in Water at 20°C
Formula Weight 32.04
Percent Purity >99.9%
Grade LC-MS
Chemical Name or Material Methanol, B&J Brand™ LC-MS, for LC-MS and HPLC, >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Methanol