missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, 99.9% min., OmniSolv™, MilliporeSigma™
Specifications
Specifications
| Melting Point | -97.8°C |
| Color | Colorless |
| Density | 0.79g/cm3 at 20°C |
| Flash Point | Closed cup: 12°C (53.6°F), Open cup: 15.85°C (60.5°F) |
| Ignition Point | 464°C (867.2°F) |
| Odor | Alcohol-like |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Assay Percent Range | 99.9%, min. |
| Linear Formula | CH3OH |
| Show More |
Safety and Handling
| DOTInformation | DOT Class: 3, Packaging Group: II |
| Recommended Storage | Room Temperature |