missing translation for 'onlineSavingsMsg'
Learn More

Methanol, 50% (v/v), Ricca Chemical

Catalog No. 482016
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
482016 500 mL
482032 1 L
48201 4 L
48205 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 482016 Supplier Ricca Chemical Company Supplier No. 482016
Only null left

Chemical Identifiers

CAS 7732-18-5 , 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) Mixture
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Color Colorless
CAS Min % 49.0
CAS Max % 51.0
Quantity 500 mL
Packaging Natural Poly Bottle
Physical Form Liquid
Boiling Point 66°C
Percent Purity 50%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 50% (v/v)
Chemical Name or Material Methanol
Show More Show Less