missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonyl chloride, 99.5%

Catalog No. AC125645000
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC125645000 500 mL Glass bottle
AC125641000 100 mL Glass bottle
AC125642500 250 mL Glass bottle
AC125640010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC125645000 Supplier Thermo Scientific Chemicals Supplier No. 125645000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 124-63-0 | CH3ClO2S | 114.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 124-63-0
Molecular Formula CH3ClO2S
Molecular Weight (g/mol) 114.54
MDL Number MFCD00007454
InChI Key QARBMVPHQWIHKH-UHFFFAOYSA-N
Synonym methanesulfuryl chloride, methyl sulfochloride, mesyl chloride, chloromethyl sulfone, chloro methyl sulfone, methanesulfonic acid chloride, methanesulphonyl chloride, mesylchloride, methylsulfonyl chloride, methyl sulfonyl chloride
PubChem CID 31297
IUPAC Name methanesulfonyl chloride
SMILES CS(Cl)(=O)=O

Specifications

Melting Point -33.0°C
Density 1.4800 g/mL
Boiling Point 161.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging Glass bottle
Linear Formula CH3SO2Cl
Refractive Index 1.4435 to 1.4455
Quantity 500 mL
Beilstein 04,5
Fieser 01,662; 02,268; 04,326; 05,435; 06,362; 07,225; 11,322; 13,176; 17,180
Specific Gravity 1.48
Merck Index 15,6027
Solubility Information Solubility aq. soln.: insoluble (reacts with water). Other solubilities: soluble in ethanol, ether, acetone and other, common organic solvents
Viscosity 1.97 mPa.s (25°C)
Formula Weight 114.55
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material Methanesulfonyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
May cause an allergic skin reaction.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Toxic if swallowed.
May cause respiratory irritation.
May be corrosive
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at
EINECSNumber 204-706-1

RUO – Research Use Only